3-(4-Bromo-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59465-500MG 59465-1G 59465-5G 59465-25G
77330-546EA 100.94 USD
77330-546 77342-944 77366-758 77378-828
3-(4-Bromo-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(4-Bromo-2-fluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrFNO₃
MW: 306.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759177
CAS Number: 2755723-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR