3-(2-Chloro-5-(trifluoromethyl)phenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%

Supplier: AOBChem USA

Warning

59460-1G 59460-5G 59460-25G
77341-802EA 104.03 USD
77341-802 77366-754 77378-824
3-(2-Chloro-5-(trifluoromethyl)phenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥95%
3-(2-Chloro-5-(trifluoromethyl)phenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClF₃NO₃
MW: 311.685 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759173
CAS Number: 2755722-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR