3-(2-Bromo-5-methoxyphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
Supplier: AOBChem USA
|
Warning
|
59459-500MG
59459-1G
59459-5G
59459-25G
77330-544EA
100.94
USD
77330-544
77342-942
77366-752
77378-822
3-(2-Bromo-5-methoxyphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(2-Bromo-5-methoxyphenyl)-3-hydroxy-N-methoxy-N-methylpropanamide
Precaución: for Research Use Only
Formula:
C₁₂H₁₆BrNO₄ MW: 318.164 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759172 CAS Number: 2755718-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...