3-(3,5-Difluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%

Supplier: AOBChem USA

Warning

59455-500MG 59455-1G 59455-5G 59455-25G
77330-542EA 100.94 USD
77330-542 77342-940 77366-750 77378-820
3-(3,5-Difluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide ≥97%
3-(3,5-Difluorophenyl)-3-hydroxy-N-methoxy-N-methylpropanamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₂NO₃
MW: 245.223 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759168
CAS Number: 2755718-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR