2-Methoxy-5-(p-tolyl)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

59444-500MG 59444-1G 59444-5G 59444-25G
77330-538EA 204.97 USD
77330-538 77354-740 77366-746 77388-440
2-Methoxy-5-(p-tolyl)pyrimidine ≥97%
2-Methoxy-5-(p-tolyl)pyrimidine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O
MW: 200.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759163
CAS Number: 27956-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR