5-(4-Fluorophenyl)-2-methoxypyrimidine ≥97%

Supplier: AOBChem USA

Warning

59443-500MG 59443-1G 59443-5G
77340-244EA 854.9 USD
77340-244 77354-738 77378-816
5-(4-Fluorophenyl)-2-methoxypyrimidine ≥97%
5-(4-Fluorophenyl)-2-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₉FN₂O
Storage Temperature: Ambient
MDL Number: MFCD34759162
CAS Number: 2764728-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR