2-Methoxy-5-phenylpyrimidine ≥97%
Supplier: AOBChem USA
|
Warning
|
59442-500MG
59442-1G
59442-5G
77340-242EA
854.9
USD
77340-242
77354-736
77378-814
2-Methoxy-5-phenylpyrimidine ≥97%
2-Methoxy-5-phenylpyrimidine
Precaución: for Research Use Only
Formula:
C₁₁H₁₀N₂O Storage Temperature: Ambient |
MDL Number:
MFCD18417358 CAS Number: 27956-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...