(2-Fluoro-4-isopropoxy-5-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

59421-500MG 59421-1G 59421-5G
77338-830EA 523.24 USD
77338-830 77354-728 77378-808
(2-Fluoro-4-isopropoxy-5-methoxyphenyl)methanol ≥97%
(2-Fluoro-4-isopropoxy-5-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₅FO₃
MW: 214.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28530202
CAS Number: 1784994-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR