2-Fluoro-4-isopropoxy-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

59420-500MG 59420-1G 59420-5G
77337-380EA 412 USD
77337-380 77354-726 77378-806
2-Fluoro-4-isopropoxy-5-methoxybenzaldehyde ≥97%
2-Fluoro-4-isopropoxy-5-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28641689
CAS Number: 1784410-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR