3-(2-Bromo-5-ethoxyphenyl)-3-hydroxypropanoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
59393-500MG
59393-1G
59393-25G
59393-5G
77330-532EA
298.7
USD
77330-532
77354-724
77388-434
77378-804
3-(2-Bromo-5-ethoxyphenyl)-3-hydroxypropanoic acid ≥97%
3-(2-Bromo-5-ethoxyphenyl)-3-hydroxypropanoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₃BrO₄ MW: 289.123 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34759133 CAS Number: 2901106-95-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...