3-(4-Bromo-2-methylphenyl)-3-hydroxypropanoic acid ≥97%

Supplier: AOBChem USA

Warning

59392-500MG 59392-1G 59392-5G
77339-224EA 568.56 USD
77339-224 77354-722 77378-802
3-(4-Bromo-2-methylphenyl)-3-hydroxypropanoic acid ≥97%
3-(4-Bromo-2-methylphenyl)-3-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MW: 259.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32118731
CAS Number: 1881664-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR