3-hydroxy-3-(4-(trifluoromethyl)phenyl)propanoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
59387-500MG
59387-1G
59387-5G
59387-25G
77330-528EA
153.47
USD
77330-528
77345-496
77366-740
77388-430
3-hydroxy-3-(4-(trifluoromethyl)phenyl)propanoic acid ≥97%
3-hydroxy-3-(4-(trifluoromethyl)phenyl)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉F₃O₃ MW: 234.172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16756900 CAS Number: 1249638-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...