4-Fluoro-3-(hydroxy(phenyl)methyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

59373-500MG 59373-1G 59373-5G 59373-25G
77330-526EA 202.91 USD
77330-526 77354-718 77366-738 77388-428
4-Fluoro-3-(hydroxy(phenyl)methyl)benzonitrile ≥97%
4-Fluoro-3-(hydroxy(phenyl)methyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₀FNO
MW: 227.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759130
CAS Number: 1499388-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR