4-Fluoro-3-((4-Fluorophenyl)(hydroxy)methyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

59372-500MG 59372-1G 59372-5G 59372-25G
77330-524EA 161.71 USD
77330-524 77345-896 77366-736 77388-426
4-Fluoro-3-((4-Fluorophenyl)(hydroxy)methyl)benzonitrile ≥97%
4-Fluoro-3-((4-Fluorophenyl)(hydroxy)methyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₉F₂NO
MW: 245.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759129
CAS Number: 1519447-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR