4-Fluoro-3-(hydroxy(thiophen-2-yl)methyl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

59371-500MG 59371-1G 59371-5G 59371-25G
77330-522EA 159.65 USD
77330-522 77345-754 77366-734 77388-424
4-Fluoro-3-(hydroxy(thiophen-2-yl)methyl)benzonitrile ≥95%
4-Fluoro-3-(hydroxy(thiophen-2-yl)methyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₂H₈FNOS
Storage Temperature: Ambient
CAS Number: 2237249-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR