1-(4-Bromo-2-fluoro-3-iodophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

59348-500MG 59348-1G 59348-5G 59348-25G
77330-520EA 116.39 USD
77330-520 77343-426 77366-732 77378-800
1-(4-Bromo-2-fluoro-3-iodophenyl)ethanone ≥97%
1-(4-Bromo-2-fluoro-3-iodophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅BrFIO
MW: 342.932 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759126
CAS Number: 2383369-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR