(3-Bromo-5-chloro-4-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59342-500MG 59342-1G 59342-5G
77335-478EA 355.35 USD
77335-478 77354-712 77378-796
(3-Bromo-5-chloro-4-fluorophenyl)(methyl)sulfane ≥97%
(3-Bromo-5-chloro-4-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrClFS
Storage Temperature: Ambient
MDL Number: MFCD34759123
CAS Number: 2710186-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR