(3-Bromo-4-fluoro-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59340-500MG 59340-1G 59340-5G
77330-516EA 345.05 USD
77330-516 77354-710 77378-794
(3-Bromo-4-fluoro-5-methylphenyl)(methyl)sulfane ≥97%
(3-Bromo-4-fluoro-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrFS
Storage Temperature: Ambient
MDL Number: MFCD34759121
CAS Number: 2706032-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR