2-Fluoro-3-formyl-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

59336-500MG 59336-1G 59336-5G
77335-862EA 367.71 USD
77335-862 77354-708 77378-792
2-Fluoro-3-formyl-5-(methylthio)benzoic acid ≥97%
2-Fluoro-3-formyl-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇FO₃S
Storage Temperature: Ambient
MDL Number: MFCD34759119
CAS Number: 2807440-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR