(2-Chloro-6-fluoro-3-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59318-1G 59318-5G 59318-25G
77341-938EA 108.15 USD
77341-938 77366-724 77378-786
(2-Chloro-6-fluoro-3-methoxyphenyl)(methyl)sulfane ≥97%
(2-Chloro-6-fluoro-3-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFOS
MW: 206.665 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759113
CAS Number: 2709834-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR