2-Chloro-4-fluoro-3-iodo-1-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

59316-500MG 59316-1G 59316-5G 59316-25G
77330-510EA 147.29 USD
77330-510 77345-220 77366-722 77388-420
2-Chloro-4-fluoro-3-iodo-1-methoxybenzene ≥95%
2-Chloro-4-fluoro-3-iodo-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34662883
CAS Number: 2385446-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR