(5-Bromo-2-fluoro-3-(methoxymethyl)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59308-500MG 59308-1G 59308-5G
77330-508EA 349.17 USD
77330-508 77354-702 77378-780
(5-Bromo-2-fluoro-3-(methoxymethyl)phenyl)(methyl)sulfane ≥97%
(5-Bromo-2-fluoro-3-(methoxymethyl)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀BrFOS
MW: 265.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759108
CAS Number: 2807442-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR