(5-Bromo-2-fluoro-3-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

59307-500MG 59307-1G 59307-5G
77330-506EA 347.11 USD
77330-506 77354-700 77378-778
(5-Bromo-2-fluoro-3-(methylthio)phenyl)methanol ≥97%
(5-Bromo-2-fluoro-3-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈BrFOS
MW: 251.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759107
CAS Number: 2764728-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR