Ethyl 5-bromo-2-fluoro-3-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

59305-500MG 59305-1G 59305-5G
77330-504EA 329.6 USD
77330-504 77354-698 77378-776
Ethyl 5-bromo-2-fluoro-3-(methylthio)benzoate ≥97%
Ethyl 5-bromo-2-fluoro-3-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₂S
Storage Temperature: Ambient
MDL Number: MFCD34759105
CAS Number: 2755718-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR