5-Bromo-2-fluoro-3-(methylthio)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

59303-500MG 59303-1G 59303-5G
77336-430EA 387.28 USD
77336-430 77354-694 77378-774
5-Bromo-2-fluoro-3-(methylthio)benzoic acid ≥95%
5-Bromo-2-fluoro-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂S
MW: 265.099 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759103
CAS Number: 2755724-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR