Methyl 3-isopropoxy-5-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

59301-500MG 59301-1G 59301-5G 59301-25G
77330-500EA 281.19 USD
77330-500 77354-692 77366-718 77388-416
Methyl 3-isopropoxy-5-(methylthio)benzoate ≥97%
Methyl 3-isopropoxy-5-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃S
MW: 240.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759101
CAS Number: 2807443-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR