3-Isopropoxy-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

59300-500MG 59300-1G 59300-5G 59300-25G
77330-498EA 251.32 USD
77330-498 77354-690 77366-716 77388-414
3-Isopropoxy-5-(methylthio)benzoic acid ≥97%
3-Isopropoxy-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃S
Storage Temperature: Ambient
MDL Number: MFCD34759100
CAS Number: 2764730-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR