(3-Bromo-5-isopropoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59296-500MG 59296-1G 59296-5G 59296-25G
77330-494EA 115.36 USD
77330-494 77343-398 77366-712 77378-772
(3-Bromo-5-isopropoxyphenyl)(methyl)sulfane ≥97%
(3-Bromo-5-isopropoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrOS
MW: 261.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759097
CAS Number: 2755723-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR