2,5-Dichloro-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

59276-500MG 59276-1G 59276-5G
77338-896EA 528.39 USD
77338-896 77354-686 77378-768
2,5-Dichloro-4-iodobenzaldehyde ≥97%
2,5-Dichloro-4-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₂IO
MW: 300.909 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759092
CAS Number: 2091672-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR