(4-Ethoxy-5-fluoro-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59261-500MG 59261-1G 59261-5G
77339-550EA 596.37 USD
77339-550 77354-682 77378-764
(4-Ethoxy-5-fluoro-2-methylphenyl)(methyl)sulfane ≥97%
(4-Ethoxy-5-fluoro-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃FOS
Storage Temperature: Ambient
MDL Number: MFCD34759087
CAS Number: 2709907-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR