1-Ethoxy-2-fluoro-4-iodo-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

59260-500MG 59260-1G 59260-5G
77336-158EA 379.04 USD
77336-158 77354-680 77378-762
1-Ethoxy-2-fluoro-4-iodo-5-methylbenzene ≥95%
1-Ethoxy-2-fluoro-4-iodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO
MW: 280.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759086
CAS Number: 2092362-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR