1-Bromo-4,5-dimethoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59259-500MG 59259-1G 59259-5G 59259-25G
77330-492EA 184.37 USD
77330-492 77347-238 77366-710 77388-410
1-Bromo-4,5-dimethoxy-2-methylbenzene ≥97%
1-Bromo-4,5-dimethoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD12095228
CAS Number: 52806-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR