5-Ethoxy-2-methoxy-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

59255-500MG 59255-1G 59255-5G
77335-936EA 369.77 USD
77335-936 77354-674 77378-756
5-Ethoxy-2-methoxy-1,3-dimethylbenzene ≥97%
5-Ethoxy-2-methoxy-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28794117
CAS Number: 1806428-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR