1-Bromo-3-(difluoromethoxy)-2-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59253-500MG 59253-1G 59253-5G
77337-726EA 438.78 USD
77337-726 77354-670 77378-754
1-Bromo-3-(difluoromethoxy)-2-fluoro-5-methylbenzene ≥97%
1-Bromo-3-(difluoromethoxy)-2-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759083
CAS Number: 2169527-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR