1-Bromo-2-fluoro-3-isopropoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59252-500MG 59252-1G 59252-5G 59252-25G
77330-486EA 210.12 USD
77330-486 77354-668 77366-706 77388-406
1-Bromo-2-fluoro-3-isopropoxy-5-methylbenzene ≥97%
1-Bromo-2-fluoro-3-isopropoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO
MW: 247.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759082
CAS Number: 2227627-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR