1-Bromo-3-ethoxy-2-fluoro-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59251-500MG 59251-1G 59251-5G 59251-25G
77330-484EA 201.88 USD
77330-484 77354-666 77366-704 77388-404
1-Bromo-3-ethoxy-2-fluoro-5-methylbenzene ≥97%
1-Bromo-3-ethoxy-2-fluoro-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759081
CAS Number: 2383609-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR