5-Bromo-2-fluoro-3-methoxy-2'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

59250-500MG 59250-1G 59250-5G
77340-140EA 791.04 USD
77340-140 77354-664 77378-752
5-Bromo-2-fluoro-3-methoxy-2'-methyl-1,1'-biphenyl ≥97%
5-Bromo-2-fluoro-3-methoxy-2'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
Storage Temperature: Ambient
MDL Number: MFCD34759080
CAS Number: 2901086-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR