5-Bromo-2-fluoro-3-methoxy-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

59249-500MG 59249-1G 59249-25G 59249-5G
77330-482EA 305.91 USD
77330-482 77354-662 77388-402 77378-750
5-Bromo-2-fluoro-3-methoxy-4'-methyl-1,1'-biphenyl ≥97%
5-Bromo-2-fluoro-3-methoxy-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO
MW: 295.147 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759079
CAS Number: 2807465-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR