Ethyl 3-chloro-4-fluoro-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

59246-500MG 59246-1G 59246-5G
77338-670EA 507.79 USD
77338-670 77354-660 77378-748
Ethyl 3-chloro-4-fluoro-5-methoxybenzoate ≥97%
Ethyl 3-chloro-4-fluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
MW: 232.636 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759076
CAS Number: 2112468-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR