(3-Chloro-4-fluoro-5-methoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

59244-500MG 59244-1G 59244-5G
77338-858EA 526.33 USD
77338-858 77354-656 77378-744
(3-Chloro-4-fluoro-5-methoxyphenyl)methanol ≥97%
(3-Chloro-4-fluoro-5-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈ClFO₂
MW: 190.599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28523091
CAS Number: 1783586-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR