2-(Benzyloxy)-4-bromo-3-chloro-1-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

59235-500MG 59235-1G 59235-25G 59235-5G
77330-478EA 301.79 USD
77330-478 77354-652 77388-398 77378-740
2-(Benzyloxy)-4-bromo-3-chloro-1-fluorobenzene ≥97%
2-(Benzyloxy)-4-bromo-3-chloro-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34759068
CAS Number: 2807467-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR