1-(2,6-Difluoro-3-methylphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

59217-500MG 59217-1G 59217-5G 59217-25G
77330-476EA 100.94 USD
77330-476 77342-938 77366-700 77378-738
1-(2,6-Difluoro-3-methylphenyl)ethanol ≥97%
1-(2,6-Difluoro-3-methylphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD24310362
CAS Number: 1173241-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR