(2,6-Difluoro-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59216-1G 59216-5G 59216-25G
77340-758EA 60.77 USD
77340-758 77366-698 77378-736
(2,6-Difluoro-3-methylphenyl)(methyl)sulfane ≥97%
(2,6-Difluoro-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈F₂S
Storage Temperature: Ambient
MDL Number: MFCD28790541
CAS Number: 1804515-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR