1,3-Difluoro-2-iodo-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59215-500MG 59215-1G 59215-5G 59215-25G
77330-474EA 211.15 USD
77330-474 77354-650 77366-696 77388-396
1,3-Difluoro-2-iodo-4-methylbenzene ≥97%
1,3-Difluoro-2-iodo-4-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂I
MW: 254.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24310274
CAS Number: 1557651-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR