4-(Benzyloxy)-3-chloro-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

59144-500MG 59144-1G 59144-5G 59144-25G
77330-464EA 116.39 USD
77330-464 77343-490 77366-688 77378-730
4-(Benzyloxy)-3-chloro-5-methylbenzoic acid ≥97%
4-(Benzyloxy)-3-chloro-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₃
Storage Temperature: Ambient
MDL Number: MFCD34759024
CAS Number: 1891969-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR