3-Chloro-4-ethoxy-5-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

59132-500MG 59132-1G 59132-5G 59132-25G
77330-462EA 281.19 USD
77330-462 77354-638 77366-686 77388-388
3-Chloro-4-ethoxy-5-methylbenzoic acid ≥95%
3-Chloro-4-ethoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
MW: 214.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759013
CAS Number: 2385198-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR