(3-Chloro-4-methoxy-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

59123-500MG 59123-1G 59123-5G
77339-548EA 596.37 USD
77339-548 77354-634 77378-724
(3-Chloro-4-methoxy-5-methylphenyl)(methyl)sulfane ≥97%
(3-Chloro-4-methoxy-5-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁ClOS
MW: 202.701 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759005
CAS Number: 2709329-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR