1-Chloro-5-iodo-2-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59122-500MG 59122-1G 59122-5G
77336-832EA 394.49 USD
77336-832 77354-632 77378-722
1-Chloro-5-iodo-2-methoxy-3-methylbenzene ≥97%
1-Chloro-5-iodo-2-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClIO
Storage Temperature: Ambient
MDL Number: MFCD34759004
CAS Number: 2383904-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR