1-(4-Isopropoxy-3-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

59121-500MG 59121-1G 59121-5G 59121-25G
77330-458EA 210.12 USD
77330-458 77354-630 77366-684 77388-384
1-(4-Isopropoxy-3-methylphenyl)ethanone ≥97%
1-(4-Isopropoxy-3-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
MDL Number: MFCD11053260
CAS Number: 105321-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR