(4-Isopropoxy-3-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

59120-500MG 59120-1G 59120-5G 59120-25G
77330-456EA 168.92 USD
77330-456 77346-288 77366-682 77388-382
(4-Isopropoxy-3-methylphenyl)(methyl)sulfane ≥95%
(4-Isopropoxy-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₆OS
MW: 196.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759003
CAS Number: 2764731-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR